missing translation for 'onlineSavingsMsg'
Learn More

ALPPL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 18506-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16848524

  • 3828.53 NOK / 150µL
Estimated Shipment: 13-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

There are at least four distinct but related alkaline phosphatases: intestinal, placental, placental-like, and liver/bone/kidney (tissue non-specific). The product of this gene is a membrane bound glycosylated enzyme, localized to testis, thymus.
Specifications

Specifications

ALPPL2
Polyclonal
Unconjugated
ALPPL2
ALP 1, ALPG, ALPPL, ALPPL2, GCAP, Germ cell alkaline phosphatase, PLAP like
Rabbit
Antigen Affinity Chromatography
RUO
11650, 251, 367308
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P10696, P24823
Alppl2
ALPPL2 Fusion Protein Ag13360
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers