missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALPPL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18506-1-AP-20UL
This item is not returnable.
View return policy
Description
There are at least four distinct but related alkaline phosphatases: intestinal, placental, placental-like, and liver/bone/kidney (tissue non-specific). The product of this gene is a membrane bound glycosylated enzyme, localized to testis, thymus.Specifications
| ALPPL2 | |
| Polyclonal | |
| Unconjugated | |
| ALPPL2 | |
| ALP 1, ALPG, ALPPL, ALPPL2, GCAP, Germ cell alkaline phosphatase, PLAP like | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11650, 251, 367308 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P10696, P24823 | |
| Alppl2 | |
| ALPPL2 Fusion Protein Ag13360 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction