Learn More
Ansell™ AlphaTec™ Solvex™37-186
Product Code. 15856932 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
11
Unit Size:
1 pair
Product Code. 15856932
Brand: Ansell™ 37186090
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
High-comfort, chemical resistant glove for a wide range of applications
  • Available in a wide selection of lengths, thicknesses, sizes and linings, the Ansell Sol-Vex™ nitrile glove is designed to deliver optimal results in wet or dry work environments where chemical resistance is crucial. Fully reusable, with an unequaled abrasion resistance, it provides superb comfort for the wearer.
  • Sol-Vex™ nitrile gloves are the ideal choice for safe handling in a wide range of work environments where harsh chemicals are present
  • The unlined version of the glove has no inner cotton flocking, reducing risk of lint contamination and making it ideal for production areas sensitive to the introduction of external impurities. 
  • The reversed lozenge finish further enhances levels of grip.
  • The sandpatch finish gives the glove a smooth surface that decreases indirect costs thanks to fewer rejects of fragile parts.
TRUSTED_SUSTAINABILITY
Specifications
Back Material Nitrile
Certifications/Compliance EN ISO 374-1:2016/Type A/AGJKLPT, EN ISO 374-5:2016 Bacteria, EN388:2016/4102X
Classification Category III
Cuff Material Nitrile
Cut Style Index
Description High-comfort, chemical resistant glove for a wide range of applications
Coating Material Nitrile
Color Green
Cut Resistance Rating 1
Disposable Reusable
Exterior Finish Sandpatch
Grade Industrial grade
Hand Specific
Latex Free Yes
Powdered Powder Free
Product Type Immersion Gloves
Size 9
Sterility Non-sterile
Style 37-186
Thickness (Metric) 0.65 mm
For Use With (Application) Chemical and Liquid Protetion
Glove Cuff Color Green
Glove Cuff Style Straight
Model 37-186
Packaging Type Polybag/cardboard box
Puncture Rating 2
Quantity 1 Pair/bag, 12 Pair/carton
Resistant To Chemical and Liquid Protection
Series Solvex™
Dimensions (L x W) 453 x 112 mm
Length (English) 453 mm
Width (Metric) 112 mm
Weight (Metric) 88.5 g