Learn More
ANSELL™ AlphaTec™ Nitrile Gloves
Product Code. 17993552
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
12 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17993552 9 12 pairs
17923522 8 12 pairs
17973552 7 12 pairs
17983552 10 12 pairs
17933552 11 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17993552 Supplier ANSELL™ Supplier No. 37300090
Only null left
Add to basket
Add to basket
All day comfort at exceptional value with these 0.20 mm thick nitrile gloves.
  • 8 mil or 0.2 mm thickness allows dexterity and tactility without a loss of durability
  • NBR offering protection against light acids, greases, oils, and liquids
  • Engineered fit to allow optimum balance of fit and flexibility both on its own or as an over glove
  • The latex-free formulation of these gloves presents no risk of Type 1 allergies
  • Long cuff extends to the forearm
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 0
Certifications/Compliance (Europe) EN 421:2010, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, Food Contact, ISO 18889:2019
Chlorinated No
Applications Industrial
Color Green
Finger Thickness (English) 7.9 mil
Finger Thickness (Metric) 0.2 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 7.9 mil
Palm Thickness (Metric) 0.2 mm
Product Type Chemical-resistant Gloves
Size 9
Thickness (English) 7.9 mil
Thickness (Metric) 0.2 mm
EN 421 Yes
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES JKT
EN ISO 374 - 5 Bacteria, Fungi WITH Virus Protection
Glove Cuff Style Gauntlet
Grip Design Lozenge
Lining Unlined
Length (English) 12.2 in.
Length (Metric) 310 mm