Learn More
Ansell™ AlphaTec™ Apron with Sleeves Ultrasonically Welded and Taped
Shop All Ansell Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- The AlphaTec™ 3000 Apron with Sleeves Ultrasonically Welded and Taped - Model 215 is made with a multi-layer fabric, offering wearers protection against several inorganic chemicals and biological hazards
- This highly visible, bright yellow chemical protective suit is designed to protect workers and enhance workplace safety
- Made from laminate fabric, the AlphaTec™ 3000 Apron with Sleeves Ultrasonically Welded Taped - Model 215 ensures comfort and durability
Specifications
Specifications
| Type | Apron |
| Size | 3X-Large |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | Yellow |
| Disposable | Single-use |
| Closure Type | Waist Ties |
| Sterility | Non-sterile |
| Latex Free | Yes |
| No. of Pockets | None |
| Seam Type | Welded and Taped |
| Show More |