Learn More
Ansell AlphaTec™ 87-137 Super Thin Unlined Latex Gloves
Product Code. 16378735 Shop All Ansell Products
Click to view available options
Size:
6.5
7.5
8.5
9.5
Unit Size:
12 pairs
Product Code. 16378735
Brand: Ansell 87137075BP
Only null left
Add to basket
Add to basket
Gloves are super thin, highly flexible, and enhance grip capabilities in wet or dry conditions.
  • Highly resistant to water based chemicals
  • Beaded cuff for tear resistance and easy donning
  • Diamond pattern on palms and fingers enhances grip in wet or dry conditions
  • Approved for use in food-safe applications
  • AQL 0.6
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) CE 0493, EN 420:2003 + A1:2009
Applications Industrial
Color Natural
Exterior Finish Fully Textured
Finger Thickness (English) 0.018 in.
Finger Thickness (Metric) 0.5 mm
Latex Free No
Material Latex
Palm Thickness (English) 0.016 in.
Palm Thickness (Metric) 0.4 mm
Product Type Latex Gloves
Size 7.5
Thickness (English) 0.018 in.
Thickness (Metric) 0.5 mm
EN388 Standard X010A
EN 421 Yes
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES KLT
Glove Cuff Style Beaded
Resistant To Water Based Chemicals
Series 87-137
Length (Metric) 320 mm