Learn More
Ansell AlphaTec™ 58-008 Extended Full Arm Coverage Gloves
Product Code. p-7241922 Shop All Ansell Products
Click to view available options
Size:
8
9
10
Unit Size:
10 pairs
Product Code. 17815641
Brand: Ansell 58008080
Only null left
Add to basket
Add to basket
Dry and wet grip enhance dexterity and reduce hand fatigue. Wide cuff to allow donning over protection clothing.
  • Nitrile polymer for excellent mechanical protection as well as resistance to oils, fats, greases and hydrocarbons
  • AlphaTec 58-008 has Type A chemical protection under new ENISO 374:2016
  • Lozenge grip for enhance grip in dry and wet conditions
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 388:2016 +A1:2018, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, EN 388:2016, EN 420:2003 + A1:2009, EN ISO 374-1:2016, EN ISO 374-5:2016, Food Contact
Chlorinated Yes
Accelerator Free No
Applications Industrial
Coating Material Nitrile
Color Green
Latex Free Yes
Material Nitrile
Product Type Protective Gloves
Size 8
Gender Unisex
Glove Cuff Style Extended
Lining Lined
Model 58-008