Learn More
Ansell™ AlphaTec™ 57-435 Series Nitrile Immersion Gloves
Product Code. 15375191 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
12 pairs
Product Code. Size unitSize
15375191 11 12 pairs
15599441 7 12 pairs
15509451 8 12 pairs
15519451 9 12 pairs
15385181 10 12 pairs
5 options
Product Code. 15375191
Brand: Ansell™ 58435110
Only null left
Add to basket
Add to basket
Provides confident grip performance with enhanced flexibility. Ansell Alphatec™ 58-435 Series Nitrile Immersion Gloves has optimal control when handling oily or wet items.
  • Chemical and liquid protection gloves
  • Features Ansell Grip Technology™ for optimal control when handling oily or wet items
  • Less force is required on slippery objects, reducing hand and arm fatigue
  • Offers improved flexibility and dexterity thanks to cotton-flocked lined nitrile shell and snug, second-skin tailoring that tightens the fit across the palm
  • Has 6-mil shell effective against hydrocarbon derivatives, alcohol bases, many solvents and esters
  • Offers superior snag, puncture and abrasion protection for long-term wear
  • Foldable gutter cuff helps prevent dripping onto the forearm, while an extra-long cuff extends protection to the upper forearm
Applications:
  • Chemical handling
  • Petrochemicals
  • Rubber and rubber additives
  • Handling fluid-coated metal parts
  • Industrial tool maintenance
  • Handling waste oils
  • Waste and water treatment
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 4
Back Material Nitrile
Chlorinated No
Cuff Material Nitrile
Applications Industrial
Color Gray
Finger Thickness (English) 0.0278 in.
Finger Thickness (Metric) 0.72 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 0.0276 in.
Palm Thickness (Metric) 0.7 mm
Product Type Immersion Gloves
Size 11
Thickness (English) 0.7 mm
Thickness (Metric) 0.0276 in.
EN 421 No
EN ISO 374-1 Type A
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Gauntlet
Grip Design Ansell Grip Technology
Liner Material Cotton Flocking
Lining Unlined
Quantity 12 Pairs
Resistant To Chemical resistance
Series 57-435