Learn More
Ansell AlphaTec™ 4000 Jacket
Product Code. p-7228388
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 20
Product Code. 17100695
Brand: Ansell GR40T0023005
Only null left
Add to basket
Add to basket
A unique multi-layer barrier fabric renowned for its lightweight, yet robust textile feel and exceptional barrier to organic & inorganic chemicals.
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Designed to protect
  • Silicone free
TRUSTED_SUSTAINABILITY
Specifications
Size X-Large
Gender Unisex
Material Multi-layer Barrier Laminate Fabric
Color Green
Closure Type Dual Zipper
Seam Type Welded and Taped
Certifications/Compliance (Europe) Partial Body Protection Only, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN 14126:2003
Product Type Chemical-resistant Jacket
Style G40T230
For Use With (Application) Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes
Weight (Metric) 103 gsm