Learn More
Ansell AlphaTec™ 4000
Product Code. 17100665
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Style:
G40T103
G40T111
G40T121
G40T125B
G40T125D
G40T132
G40T151
G40T151B
G40T162
Unit Size:
Pack of 10
Pack of 12
Pack of 8
Product Code. Size Style unitSize
17100665 3X-Large G40T125D Pack of 8
17110645 X-Small G40T111 Pack of 10
17120655 Medium G40T125D Pack of 8
17160635 Medium G40T103 Pack of 10
17170635 Large G40T103 Pack of 10
17180635 X-Large G40T103 Pack of 10
17190635 2X-Large G40T103 Pack of 10
17100645 3X-Large G40T103 Pack of 10
17190645 Small G40T125B Pack of 8
17110655 Medium G40T125B Pack of 8
17130655 Large G40T125B Pack of 8
17150655 X-Large G40T125B Pack of 8
17170655 2X-Large G40T125B Pack of 8
17190655 3X-Large G40T125B Pack of 8
17100655 Small G40T125D Pack of 8
17120645 Small G40T121 Pack of 10
17140655 Large G40T125D Pack of 8
17160655 X-Large G40T125D Pack of 8
17180655 2X-Large G40T125D Pack of 8
17130665 Small G40T132 Pack of 12
17140665 Medium G40T132 Pack of 12
17150665 Large G40T132 Pack of 12
17160665 X-Large G40T132 Pack of 12
17170665 2X-Large G40T132 Pack of 12
17180665 3X-Large G40T132 Pack of 12
17100675 Medium G40T151 Pack of 8
17110675 Large G40T151 Pack of 8
17120675 X-Large G40T151 Pack of 8
17130675 2X-Large G40T151 Pack of 8
17140675 3X-Large G40T151 Pack of 8
17190665 Small G40T151B Pack of 8
17150675 Small G40T162 Pack of 10
17160675 Medium G40T162 Pack of 10
17170675 Large G40T162 Pack of 10
17180675 X-Large G40T162 Pack of 10
17190675 2X-Large G40T162 Pack of 10
17100685 3X-Large G40T162 Pack of 10
37 options
Product Code. 17100665
Brand: Ansell GR40T0012507G05
Only null left
Add to basket
Add to basket
A chemical protective suit that is engineered to provide an exceptional barrier against a wide range of organic and inorganic chemicals and biological agents.
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Designed to protect, typical innovative features include dual zip systems and double cuffs
  • Welded and taped seams, our highest barrier to liquids and particulates
  • Enhanced comfort, knitted inner cuffs and inner textile type fabric improves wearer acceptance
  • Anti-static
TRUSTED_SUSTAINABILITY
Specifications
Size 3X-Large
Material Multi-layer Barrier Laminate Fabric
Color Green
Seam Type Welded, Taped
Certifications/Compliance (Europe) EN 374-5:2003, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 1073-2:2002, EN 1149-5:2008, EN 14126:2003, NFPA 1992
For Use With (Application) Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Handling, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes
Style G40T125D