Learn More
Ansell AlphaTec™ 38-560 Chemical Resistant Butyl Gloves - Gauntlet Cuff
Product Code. p-7215034 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
11
Unit Size:
10 pairs
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 16845220
Brand: Ansell 38560070
Only null left
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Gloves are constructed of a thick butyl material to provide a high level of chemical resistance against agressive chemicals, without sacrificing comfort or dexterity.
  • High protection level against gases, particularly ozone
  • Extended gauntlet style cuff for additional protection
  • Beaded cuff for tear resistance and easy donning
  • Rough finish on the hand for safer handling
  • Designed for comfort with a natural, curved ergonomic shape
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Applications Industrial
Color Black
Latex Free Yes
Material Butyl Polymer
Product Type Butyl Gloves
Size 7
EN388 Standard 1020X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES ABIKPT
EN ISO 374 - 5 Yes
Glove Cuff Style Gauntlet
Resistant To Solvents, and Strong Acids
Series 38-560