Learn More
Ansell AlphaTec™ 37-340 Chemical and Heat Resistant Nitrile Gloves, Lined
Shop All Ansell Products
Click to view available options
Size:
6.5
7.5
8.5
9.5
10.5
Unit Size:
12 pairs
Description
- Very good abrasion resistance.
- Allows contact up to 100°C.
- Superior raised grip pattern for safety handlings.
- Open cuff with textile edge binding allows ventilation to keep hand cool.
Specifications
Specifications
| Color | Green |
| Material | Nitrile |
| Size | 9.5 |
| EN 421 | Yes |
| EN ISO 374-1 | Type A |
| Glove Cuff Style | Straight |
| Lining | Lined |