Learn More
Ansell AlphaTec™ 37-210 Reusable Nitrile Gloves with UltraGrip™ Technology
Product Code. 16815210 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
11
12
Unit Size:
12 pairs
Product Code. 16815210
Brand: Ansell 37210120
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
UltraGrip™ Technology delivers an innovative raised diamond grip on the glove's surface for enhanced grip capabilities in wet and oily conditions.
  • Reusable nitrile gloves designed specifically for food processing and handling
  • Raised diamond-pattern enhances grip in wet and oily conditions
  • Gloves maintain flexibility and dexterity, even at low temperatures
  • Use alone or as outer glove over thermal liners or cut-resistant gloves
  • Latex-free formulation presents no risk of Type 1 allergies
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Color Blue
Disposable Reusable
Hand Specific
Latex Free Yes
Material Nitrile
Size 12
Thickness (English) 8 mil
EN388 Standard 3001X
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES JKT
Series 37-210