Learn More
Ansell AlphaTec™ 3000 AVANT2 AIRline
Description
- Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
- Panoramic visor design ensures a good field of vision for the wearer.
- HEPA filter provides secondary protection from airline contamination and a SMC silencer ensures the noise inside the suit is always below 70 dB (even at maximum airflow).
- Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer.
Spécification
Spécification
| Color | Yellow |
| For Use With (Application) | Transferring Liquids and Solids Between Vessels and Tanks and Process Equipment, Opening and Draining Pumps Valves or Lines, Filling, Blending and Charging of Raw Materials, Mixing or Loading Chemicals, Cleaning of Plant and Machines, Unexpected Leakages Spills or Other Releases, Loading and Unloading Trucks and Vehicles, Product Inspection |
| Hood Type | Fully Encapsulated |
| Material | Multi-layer Barrier Laminate Fabric |
| Size | Large |
| Product Type | Encapsulated chemical protective suits for use with air-fed/supplied systems |
| Seam Type | Welded and Taped |
| Style | Y300757D |
| Weight (Metric) | 80 gsm |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?