missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec 3000 AVANT AIRline
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Style:
Y30T755
Y30T755D
Unit Size:
Pack of 4
Description
- Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
- Panoramic visor design ensures a good field of vision for the wearer
- A range of sleeve options available, including double cuffs and permanently attached chemical protective gloves.
- HEPA filter provides secondary protection from airline contamination and a SMC silencer ensures the noise inside the suit is always below 70 dB (even at maximum airflow).
- Suit/Belt mounted regulator complete with a low flow warning whistle the AVANT STS regulator can be mounted on the suit and removed for reuse providing cleaning and decontamination is permissible.
- Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer
- Attached socks with elasticated boot overflap - Socks are designed to be worn inside chemical protective boots with the leg overflap worn outside to reduce the potential for chemical ingress.
Specifications
Specifications
| Color | Yellow |
| For Use With (Application) | Air-fed/Supplied Systems |
| Hood Type | Fully Encapsulated |
| Material | Multi-layer Barrier Laminate Fabric |
| Size | X-Large |
| Product Type | Encapsulated chemical protective suits for use with air-fed/supplied systems |
| Seam Type | Welded and Taped |
| Style | Y30T755 |
| Weight (Metric) | 80 gsm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction