missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2300 COMFORT Bound
Click to view available options
:
Size: Small, Style: W23B129
Description
- An excellent barrier to resins and fibres
- Hood, arms, legs and front torso in AlphaTec™ 2300 fabric li>Air and water vapor permeable (breathable) back panel ventilates the suit, to help reduce the risk of heat stress
- Innovative fabric construction ensures that there is no risk of delamination should a wearer make contact with a tacky surface
- Anti-static
- Silicone Free, critical in spray painting applications
Specifications
Specifications
| Size | Small |
| Material | Microporous Polyethylene Laminate Non-Woven (Hood,Arms and Legs) and Polypropylene Non-Woven with Anti-Static Treatment (Back Panel) |
| Color | White, Yellow |
| Seam Type | Bound |
| Certifications/Compliance (Europe) | EN 374:2003, , EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN 1149-5:2008 |
| For Use With (Application) | Equipment Maintenance and Repair, Equipment Repair and Maintenance, Handling Structural and Body Components, Assembly & Inspection of Components, Fibre Glass/Resin Applications/Ceramic Fibres |
| Weight (Metric) | Polyethylene on bicomponent Polypropylene/Polyethylene Nonwoven 62 gsm Polypropylene nonwoven with anti-static treatment 48 gsm |
| Style | W23B129 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction