Learn More
Ansell AlphaTec™ 1800 COMFORT Bound - Model 195
Product Code. p-7228381
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 40
Pack of 50
Product Code. 16844900
Brand: Ansell WN18B0019502
Only null left
Add to basket
Add to basket
Lightweight coveralls with breathable microporous laminate technology and Type 5/6 protection.
  • 3 piece hood
  • Elasticated hood, wrists, waist and ankles
  • 2-way front zipper with resealable storm flap
  • Finger Loops
TRUSTED_SUSTAINABILITY
Specifications
Size Small
Material Microporous Non-woven Polypropylene and Polyethylene Laminate
Color White, Blue
Seam Type Bound
Coating Anti-Static Treatment (Back, Underarm and Hood)
Certifications/Compliance (Europe) EN 374:2003, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN 1149-5:2008, US Ergo Certification
For Use With (Application) Clean Room Cleaning and Preparing, Body Painting Including Surface Inspection, Production Line Support & Maintenance, Cleaning of Plant and Machines, Maintenance of Plant and Machines, Fibre Glass Resin Applications, Handling of Powders, General Purpose Auto Aftermarket
Style W18B195