Learn More
Ansell AlphaTec™ 1500 STANDARD Stitched, Model 138, Navy
Product Code. 18030120 Shop All Ansell Products
Change view
Click to view available options
Size:
4X-Large
5X-Large
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
18030120 5X-Large Each
18050120 4X-Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18030120 Supplier Ansell Supplier No. NV15S0013809
Estimated Shipment: 15-12-2026
Provides comfortable protection from hazardous particulates in general industrial environments.
SUIT FEATURES:
  • 3 piece hood
  • Elasticated hood, wrists, waist and ankles
  • 2-way front zipper with resealable storm flap
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hooded Coveralls
Size 5X-Large
Gender Unisex
Material SMS Fabric
Color Navy
Disposable Reusable
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type 2-way Zipper with Storm Flap
Seam Type Stitched
Sterility Non-sterile
Protection Class CE CATEGORY III (Type 5, Type 6)
Resistance Chemicals, Particle
Collar Type Elasticated hood
Certifications/Compliance (Europe) CE Number|EN ISO 13688:2013|EN ISO 13982-1:2004 + A1:2010|EN ISO 13982-1:2004 + A1:2010|EN ISO 13982-1:2004 + A1:2010|EN 13034:2005 + A1:2009|EN 1073-2:2002
Latex Free Yes