Learn More
Ansell AlphaTec™ 1500 PLUS Stitched Coveralls, Model 113, Orange
Product Code. p-200022875 Shop All Ansell Products
Click to view available options
Size:
6X-Large
7X-Large
Unit Size:
Each
Product Code. 18000130
Brand: Ansell OR15S0011310
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provides comfortable protection from hazardous particulates in critical environments.
DESIGN:
  • SMS fabrics are a particularly good barrier against particulates such as asbestos, brick dust and cement dust, and will provide protection from light aerosol mists; as found in some paint spray environments.
  • Anti-static suit, used for the stripping, clear up or handling of asbestos, general maintenance, construction and contract cleaning.
  • Stitched seams
  • Combining strength with particle barrier
SUIT FEATURES:
  • 3-piece hood
  • Elasticated hood, wrists, waist and ankles
  • 2-way front zipper with resealable storm flap
  • Finger loops
  • Anti-static treatment
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hooded Coveralls
Size 6X-Large
Gender Unisex
Material Anti-static SMS
Color Orange
Disposable Reusable
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic, Finger Loops
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type 2-way Zipper with Storm Flap
Seam Type Stitched
Sterility Non-sterile
Protection Class CE CATEGORY III (Type 5, Type 6)
Resistance Anti-static, Chemicals, Particle
Collar Type 3-piece hood
Certifications/Compliance (Europe) EN 374:2003|EN ISO 13982-1:2004 + A1:2010|EN 13034:2005 + A1:2009|CE Number|EN 1073-2:2002|EN 1149-5:2018|EN ISO 13688:2013
Latex Free Yes
For Use With (Application) Unpacking of bulk goods, Loading and unloading of proces equipment, Body Surface checking, marking and touching up, Assembly and inspection of components, Painting or cleaning tools or robots, Production line support and maintenance