missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Skytec™ Alphachem X350 Disposable Coverall
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 10
Description
- Manufactured using polyethylene and EVOH barrier composite, bonded structural spun-bonded and polypropylene base layer suitable for higher-risk applications
- Tough and durable
- Improved fit, enhanced freedom of movement with a reinforced crotch
- Double zip system and grab tag to support with an emergency
- Easy to hold when wearing gloves
- Reinforced knee patches for extra durability when kneeling
- Stitched and fully taped seam stitching and elastication
- Elasticated wrists and ankles for secure fit
- Three-piece hood construction for better fit
- Available in a wide range of sizes
- Double sleeve system with inner thumb loop ensuring arm protection to support when arm is raised
Specifications
Specifications
| Product Type | Chemical-resistant Coveralls |
| Size | 2X-Large |
| Gender | Unisex |
| Material | Composite Fabric |
| Color | Orange |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction