missing translation for 'onlineSavingsMsg'
Learn More

alpha-Smooth Muscle Actin Antibody (4A4), Novus Biologicals™

Product Code. 18117091 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
Unit Size:
0.1mL
25µL
Product Code. Quantity unitSize
18117091 0.025 mL 25µL
18729883 - 0.1mL
2 options
This item is not returnable. View return policy

Product Code. 18117091

Brand: Novus Biologicals NBP222120SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 5 publications

alpha-Smooth Muscle Actin Monoclonal specifically detects alpha-Smooth Muscle Actin in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-Smooth Muscle Actin
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4A4
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000
Formulation Ascites with 0.03% Sodium Azide
Gene Accession No. P62736
Gene Alias AAT6, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, sma, smooth muscle actin
Gene Symbols ACTA2
Host Species Mouse
Immunogen Alpha-Smooth Muscle Actin Antibody (4A4) is made to a synthesized peptide of human ACTA2.
Molecular Weight of Antigen 42 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Developmental Biology, Extracellular Matrix, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 59
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.