Learn More
Alpha-Ionone (>90%), TRC
Product Code. 30389911
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30389911 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30389911 Supplier Toronto Research Chemicals Supplier No. TRCI73127510G
 Restricted Item
Estimated Shipment: 30-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material alpha-Ionone
CAS 127-41-3
Molecular Formula C13 H20 O
Quantity 10 g
Synonym (E)-alpha-Ionone, 3-Buten-2-one, 4-(2,6,6-trimethyl-2-cyclohexen-1-yl)-, (3E)-, 3-Buten-2-one, 4-(2,6,6-trimethyl-2-cyclohexen-1-yl)-, (E)- (8CI), (3E)-4-(2,6,6-Trimethyl-2-cyclohexen-1-yl)-3-buten-2-one, (5E)-Ionone, (E)-4-(2,6,6-Trimethyl-2-cyclohexen-1-yl)-3-buten-2-one, (E)-α-Ionone, (±)-trans-α-Ionone, (±)-α-Ionone, 4-(2,6,6-Trimethyl-2-cyclohexen-1-yl)-3-buten-2-one, 4-(2,6,6-Trimethyl-2-cyclohexenyl)-3-buten-2-one, trans-α-Ionone, α-Cyclocitrylideneacetone, α-Ionone
InChI Formula InChI=1S/C13H20O/c1-10-6-5-9-13(3,4)12(10)8-7-11(2)14/h6-8,12H,5,9H2,1-4H3/b8-7+
SMILES CC(=O)\C=C\C1C(=CCCC1(C)C)C
IUPAC Name (E)-4-(2,6,6-trimethylcyclohex-2-en-1-yl)but-3-en-2-one
Molecular Weight (g/mol) 192.3
Formula Weight 192.1514