missing translation for 'onlineSavingsMsg'
Learn More

alpha-Hederin, MedChemExpress

Product Code. p-200097681
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. Quantity unitSize
16471358 5 mg 5mg
16458918 10 mg 10mg
2 options
This item is not returnable. View return policy

Product Code. 16471358

Brand: Medchem Express HYN0255

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress alpha-Hederin (α-Hederin), a monodesmosidic triterpenoid saponin, exhibits promising antitumor potential against a variety of human cancer cell lines. alpha-Hederin could inhibit the proliferation and induce apoptosis of gastric cancer accompanied by glutathione decrement and reactive oxygen species generation via activating mitochondrial dependent pathway.

alpha-Hederin (α-Hederin), a monodesmosidic triterpenoid saponin, exhibits promising antitumor potential against a variety of human cancer cell lines. alpha-Hederin could inhibit the proliferation and induce apoptosis of gastric cancer accompanied by glutathione decrement and reactive oxygen species generation via activating mitochondrial dependent pathway.

Specifications

Chemical Name or Material alpha-Hederin
CAS 27013-91-8
Color White
Purity Grade Notes Research
Molecular Formula C41H66O12
Quantity 5 mg
Synonym α-Hederin
Solubility Information DMSO : ≥ 100 mg/mL (133.16 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC[C@@]1(C)[C@]2([H])CC[C@@]3(C)[C@]4(C)CC[C@@]5(C(O)=O)CCC(C)(C)C[C@@]5([H])C4=CC[C@]3([H])[C@@]2(C)CC[C@@H]1O[C@]6([H])OC[C@H](O)[C@H](O)[C@H]6O[C@@]7([H])[C@H](O)[C@H](O)[C@@H](O)[C@H](C)O7
Molecular Weight (g/mol) 750.96
Formula Weight 750.96
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.