Learn More
alpha-Benzoin oxime, 98+%
Product Code. 11403354
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. Quantity unitSize
11403354 500 g 500g
11483344 25 g 25g
11493344 100 g 100g
3 options
Product Code. 11403354
Brand: Thermo Scientific Alfa Aesar A10630.36
Request Bulk or Custom Format
CAS: 441-38-3 | C14H13NO2 | 227.26 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 441-38-3
Molecular Formula C14H13NO2
Molecular Weight (g/mol) 227.26
MDL Number MFCD00004501
InChI Key WAKHLWOJMHVUJC-FYWRMAATNA-N
Synonym 2-hydroxy-1,2-diphenylethanone oxime, e,alphar-benzoinoxime, 2r-2-hydroxy-1,2-diphenylethanone oxime, 1r,2z-2-hydroxyimino-1,2-diphenylethanol, 1r,2z-2-hydroxyimino-1,2-diphenyl-ethanol, unii-ptc1433kj8 component
PubChem CID 7057888
IUPAC Name (1R,2Z)-2-hydroxyimino-1,2-diphenylethanol
SMILES O\N=C(\C(O)C1=CC=CC=C1)C1=CC=CC=C1
Specifications
Melting Point 151°C to 154°C
Quantity 500 g
Beilstein 2051941
Sensitivity Light sensitive
Merck Index 14,1094
Formula Weight 227.26
Percent Purity ≥98%
Chemical Name or Material alpha-Benzoin oxime