Learn More
Aloxistatin, MedChemExpress
Product Code. 16482858
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Product Code. 16482858
Brand: Medchem Express HY100229
MedChemExpress Aloxistatin (E64d) is a cell-permeable and irreversible broad-spectrum cysteine protease inhibitor. Aloxistatin (E64d) exhibits entry-blocking effect for MERS-CoV.
Aloxistatin (E64d) is a cell-permeable and irreversible broad-spectrum cysteine protease inhibitor. Aloxistatin (E64d) exhibits entry-blocking effect for MERS-CoV.
Specifications
Chemical Name or Material Aloxistatin
Physical Form Solid
CAS 88321-09-9
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C17H30N2O5
For Use With (Application) Neuroscience-Neurodegeneration
Synonym E64d E64c ethyl ester
Solubility Information DMSO : 250 mg/mL (730.08 mM; Need ultrasonic) ∣Ethanol : ≥ 33.33 mg/mL (97.33 mM)
SMILES O=C([C@H]1O[C@@H]1C(N[C@H](C(NCCC(C)C)=O)CC(C)C)=O)OCC
Molecular Weight (g/mol) 342.43
Formula Weight 342.43
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.