missing translation for 'onlineSavingsMsg'
Learn More

Almorexant hydrochloride, MedChemExpress

Product Code. p-200098577
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16486877

Brand: Medchem Express HY10805A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Almorexant (ACT 078573) hydrochloride is an orally active, potent and competitive dual orexin receptor antagonist, with Kd values of 1.3 nM (OX1) and 0.17 nM (OX2), respectively.

Almorexant hydrochloride reversibly blocks signaling of orexin-A and orexin-B peptides. Almorexant hydrochloride totally blocked the intracellular Ca2+ signal pathway. Almorexant hydrochloride stimulates caspase-3 activity in AsPC-1 cells and induces apoptosis.

Specifications

Chemical Name or Material Almorexant hydrochloride
CAS 913358-93-7
Color White
Purity Grade Notes Research
Molecular Formula C29H32ClF3N2O3
Quantity 5 mg
Synonym ACT 078573 hydrochloride
Solubility Information DMSO : ≥ 46 mg/mL (83.79 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES CNC([C@@H](C1=CC=CC=C1)N2CCC3=CC(OC)=C(OC)C=C3[C@@H]2CCC4=CC=C(C(F)(F)F)C=C4)=O.Cl
Molecular Weight (g/mol) 549.02
Formula Weight 549.02
Percent Purity 99.28%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.