missing translation for 'onlineSavingsMsg'
Learn More

Allyltriphenylphosphonium chloride, 97%, Thermo Scientific™

Product Code. p-52183 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10232662 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10232662 Supplier Thermo Scientific Acros Supplier No. 352360050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18480-23-4
Molecular Formula C21H20ClP
Molecular Weight (g/mol) 338.81
MDL Number MFCD00031542
InChI Key FKMJROWWQOJRJX-UHFFFAOYSA-M
Synonym allyltriphenylphosphonium chloride, allyl triphenylphosphonium chloride, triphenyl prop-2-enyl phosphanium chloride, triphenyl prop-2-en-1-yl phosphanium chloride, acmc-1buz2, allyltriphenylphosphoniumchloride, triphenyl prop-2-enyl phosphonium chloride, phosphonium,triphenyl-2-propen-1-yl-, chloride 1:1
PubChem CID 2734120
IUPAC Name triphenyl(prop-2-en-1-yl)phosphanium chloride
SMILES [Cl-].C=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 226°C to 230°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula H2CCHCH2P(C6H5)3Cl
Quantity 5 g
Formula Weight 338.82
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material Allyltriphenylphosphonium chloride
Show More Show Less
compliance-icons
Product Identifier
  • Allyl triphenylphosphonium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.