Learn More
Allyl-d5 Bromide, TRC
Product Code. 30326299
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326299 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30326299 Supplier Toronto Research Chemicals Supplier No. TRCA549657250MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Allyl-d5 Bromide
CAS 102910-37-2
Molecular Formula C3 2H5 Br
Quantity 250 mg
Synonym 1-Propene-1,1,2,3,3-d5, 3-bromo- (9CI), Propene-1,1,2,3,3-d5, bromo- (6CI), 3-Bromo-1,1,2,3,3-pentadeuterioprop-1-ene, Allyl bromide-d5, Allyl-d5 bromide, Allyl Bromide-D5, 3-Bromo-1-propene-1,1,2,3,3-d5
InChI Formula InChI=1S/C3H5Br/c1-2-3-4/h2H,1,3H2/i1D2,2D,3D2
SMILES [2H]C(=C([2H])C([2H])([2H])Br)[2H]
IUPAC Name 3-bromo-1,1,2,3,3-pentadeuterioprop-1-ene
Molecular Weight (g/mol) 126.01
Formula Weight 124.9888