missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, Ni/Cr/Mo, AISI 8740, G87400, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Alloy Steel, Ni/Cr/Mo, AISI 8740, G87400, 100g
Specifications
Specifications
| Chemical Name or Material | Boron/Lead/Zinc/Titanium/Niobium/Oxygen/Zirconium/Vanadium/Antimony/Tungsten/Arsenic/Nitrogen/Phosphorus/Cobalt/Tin/Aluminium/Molybdenum/Silicon/Copper/Carbon/Nickel/Chromium/Manganese/Selenium/Tantalum/Magnesium/Bismuth/Calcium |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Boron 0.0002 %/Lead 0.0004 %/Zinc 0.0005 %/Titanium 0.0012 %/Niobium 0.0013 %/Oxygen 0.0013 %/Zirconium 0.0013 %/Vanadium 0.0022 %/Antimony 0.0024 %/Tungsten 0.004 %/Arsenic 0.0053 %/Nitrogen 0.0068 %/Phosphorus 0.0075 %/Cobalt 0.0078 %/Tin 0.012 %/Aluminium 0.024 %/Molybdenum 0.204 %/Silicon 0.256 %/Copper 0.27 %/Carbon 0.423 %/Nickel 0.424 %/Chromium 0.468 %/Manganese 0.842 %/Selenium (<0.003) %/Tantalum (<0.005) %/Magnesium (0.0002) %/Bismuth (0.0005) %/Calcium (0.001) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction