Learn More
Karl Hecht Allihn cooler with cut, DURAN 25cm
Brand:  Karl Hecht 41987024
Allihn cooler with cut, DURAN 25cm
Product Code. 10013701
  • ONLINE EXCLUSIVE PRICE
    1000.65 NOK
    742.38 NOK / Each
    Save 258.27 NOK
    25% Off
Only null left
Estimated Shipment: 10-03-2026