Learn More
Brady™ All Weather Vinyl Labels for M710 and BMP71
Product Code. p-7251004
Change view
Click to view available options
Color:
Tan
White
Yellow
Unit Size:
Each
Product Code. Color unitSize
17839194 White Each
17839004 Tan Each
17859064 Yellow Each
3 options
Product Code. 17839194
Brand: Brady™ 173205
Only null left
Add to basket
Add to basket
All-Weather Vinyl Labels for use with Brady™ Printers M710 and BMP71.
  • Durable, low-shrink vinyl with ultra-aggressive adhesive is ideal for labelling pipes, equipment, shelves and nearly any other industrial surface
  • Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty and harsh environment surfaces--including outdoors
  • Conforms and adheres to 'difficult' surfaces like painted cinder blocks, painted wood, textured plastic, paper-jacketed pipes and blow-molded cases
  • Resistant to exposure to chemicals, greases and oils, salt spray, cleaners and detergents and general industrial grime
  • 8 to 10 year average outdoor durability in temperatures from -40°C to +82°C (-40°F to +180°F);can be applied as low as -18°C (0°F)
  • Brady™ authentic materials work seamlessly with Brady™ printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability
  • A ribbon is required to print this material
Specifications
Certifications/Compliance ASTM D 2979, ASTM D 523
Adhesive Type Permanent Acrylic
Coating None
Color White
Finish Gloss
Format Label
Material Vinyl
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Outdoor Labels
Sterility Non-sterile
For Use With (Application) Component and Equipment Labeling, Electrical Labeling, Facility Identification, Floor Marking, Laboratory Labeling, Lean and 5S Labeling, Pipe and Valve Marking, Warehouse Marking
For Use With (Equipment) Brady™ Printers M710, BMP71
Labels per Row 1 Label
Min. Operating Temperature (Metric) -40°C
Quantity 1 Roll
Temperature Range (English) -40°F to +180°F
Temperature Range (Metric) -40°C to +82°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Dimensions (LxW) 15.24 m x 12.7 mm
Length (English) 50 ft.
Length (Metric) 15.24 m
Width (English) 0.5 in.
Width (Metric) 12.7 mm
Width (English) Printable Area 12.7 mm
Alternate Part Number M7C-500-595-WT