Learn More
S Murray™ Samco™ All Glass Luer-Tip Syringe
Product Code. 12389209
Change view
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
20 mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12389209 10 mL Each
12359209 1 mL Each
12399209 20 mL Each
12369209 2 mL Each
12379209 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12389209 Supplier S Murray™ Supplier No. G811/04
  • ONLINE EXCLUSIVE PRICE
    54.30 NOK
    41.19 NOK / Pack of 1
    Save 13.11 NOK
    24% Off
Estimated Shipment: 08-01-2027
Lägg i varukorgen
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy
Use these syringes for a variety of research applications. S Murray™ All Glass Luer-Tip Syringes are graduated and feature glass luer fittings.
  • Syringe with glass luer fitting
  • Fully interchangeable
Specifikationer
Certifications/Compliance CE
Material Glass
Product Type All Glass Luer-tip Syringe
Quantity 1/Ea.
Volume (Metric) 10 mL
Connection Type Luer Tip
Graduated Yes
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.