Learn More
Linden™ Alkaline Button Batteries
Product Code. 12737565
Change view
Click to view available options
Amperage:
44 mAh capacity
70 mAh capacity
Unit Size:
Each
Product Code. Amperage unitSize
12737565 44 mAh capacity Each
12717565 70 mAh capacity Each
2 options
Product Code. 12737565
Brand: Linden™ HGP189CD10
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Providing long shelf life for general laboratory purposes. Linden™ Alkaline Button Batteries have a leak-resistant construction.
  • Available in a variety of different voltages
  • In button form
  • High energy density
Specifications
Product Type Alkaline Battery
Quantity 1 Ea.
Voltage 1.5 V
Amperage 44 mAh capacity