Learn More
Linden™ Alkaline Button Batteries
Product Code. 12717565
Change view
Click to view available options
Amperage:
44 mAh capacity
70 mAh capacity
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Amperage Unit Size
12717565 70 mAh capacity Each
12737565 44 mAh capacity Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 12717565 Supplier Linden™ Supplier No. HGP186CD10
Estimated Shipment: 05-01-2027
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Providing long shelf life for general laboratory purposes. Linden™ Alkaline Button Batteries have a leak-resistant construction.
  • Available in a variety of different voltages
  • In button form
  • High energy density
Specifications
Product Type Alkaline Battery
Quantity 1 Ea.
Voltage 1.5 V
Amperage 70 mAh capacity