Learn More
Alimemazine hemitartrate, MedChemExpress
Product Code. 16465318
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16465318
Brand: Medchem Express HY12752A
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Alimemazine hemitartrate is a phenothiazine derivative that is generally used as an antipruritic agent and also a hemagglutinin (HA)-receptor antagonist.
Alimemazine hemitartrate is a phenothiazine derivative that is generally used as an antipruritic agent and also a hemagglutinin (HA)-receptor antagonist.
Specifications
Chemical Name or Material Alimemazine hemitartrate
Physical Form Solid
CAS 4330-99-8
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C18H22N2S.1/2C4H6O6
For Use With (Application) COVID-19-immunoregulation
Synonym Trimeprazine hemitartrate
Solubility Information H2O : 100 mg/mL (267.74 mM; Need ultrasonic) ∣DMSO : 30 mg/mL (80.32 mM; Need ultrasonic and warming)
SMILES CC(CN(C)C)CN1C2=CC=CC=C2SC3=CC=CC=C13.O=C(O)[C@H](O)[C@@H](O)C(O)=O.[0.5]
Molecular Weight (g/mol) 373.49
Formula Weight 373.49
Percent Purity 98.46%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.