missing translation for 'onlineSavingsMsg'
Learn More

Alimemazine hemitartrate, MedChemExpress

Product Code. 16411527
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16411527

Brand: Medchem Express HY12752A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Alimemazine hemitartrate is a phenothiazine derivative that is generally used as an antipruritic agent and also a hemagglutinin (HA)-receptor antagonist.

Alimemazine hemitartrate is a phenothiazine derivative that is generally used as an antipruritic agent and also a hemagglutinin (HA)-receptor antagonist.

Specifications

Chemical Name or Material Alimemazine hemitartrate
Physical Form Solid
CAS 4330-99-8
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C18H22N2S.1/2C4H6O6
For Use With (Application) COVID-19-immunoregulation
Synonym Trimeprazine hemitartrate
Solubility Information H2O : 100 mg/mL (267.74 mM; Need ultrasonic) ∣DMSO : 30 mg/mL (80.32 mM; Need ultrasonic and warming)
SMILES CC(CN(C)C)CN1C2=CC=CC=C2SC3=CC=CC=C13.O=C(O)[C@H](O)[C@@H](O)C(O)=O.[0.5]
Molecular Weight (g/mol) 373.49
Formula Weight 373.49
Percent Purity 98.46%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.