missing translation for 'onlineSavingsMsg'
Learn More

Alginic Acid, MP Biomedicals

Catalog Number p-3572852
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
This item is not returnable. View return policy
9005-32-7
(C6H8O7)A(C6H8O7)B
2r,3r,4r,5s,6s-3-2r,3s,4r,5s,6s-6-carboxy-3,4-dihydroxy-5-methoxyoxan-2-yl oxy-4,5-dihydroxy-6-methyloxane-2-carboxylic acid
This item is not returnable. View return policy
9005-32-7
(C6H8O7)A(C6H8O7)B
2r,3r,4r,5s,6s-3-2r,3s,4r,5s,6s-6-carboxy-3,4-dihydroxy-5-methoxyoxan-2-yl oxy-4,5-dihydroxy-6-methyloxane-2-carboxylic acid
Color White to Yellow
Assay Percent Range >98%
Quantity 100 g
Physical Form Powder
Chemical Name or Material Alginic Acid, Source: kelp
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'