missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALG5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16046-1-AP-20UL
This item is not returnable.
View return policy
Description
Dolichyl-phosphate beta-glucosyltransferase (ALG5), belongs to the family of glycosyltransferases, specifically the hexosyltransferases. ALG5 is widely expressed in pancreas, placenta, liver, heart, brain, kidney, skeletal muscle, and lung. ALG5 can catalyze the chemical reaction between UDP-glucose and UDP with dolichyl phosphate.Specifications
| ALG5 | |
| Polyclonal | |
| Unconjugated | |
| ALG5 | |
| ALG5, DolP glucosyltransferase, RP11 421P11.2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 295051, 29880, 66248 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9DB25, Q9Y673 | |
| Alg5 | |
| ALG5 Fusion Protein Ag8990 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction