Learn More
Alfacalcidol-d6, MedChemExpress
Description
Alfacalcidol-d6 is the deuterium labeled Alfacalcidol. Alfacalcidol is a non-selective VDR activator.
Specifications
Specifications
| Chemical Name or Material | Alfacalcidol-d6 |
| CAS | 1641940-94-4 |
| Purity Grade Notes | Research |
| Physical Form | Solid |
| Molecular Formula | C27H38D6O2 |
| Color | White |
| SMILES | O[C@H](C/1=C)C[C@H](O)CC1=C\C=C2[C@@](CC[C@@H]3[C@@H](CCCC(C([2H])([2H])[2H])C([2H])([2H])[2H])C)([H])[C@]3(C)CCC\2 |
| Industry Type | Metabolism-sugar/lipid metabolism |
| Molecular Weight (g/mol) | 406.67 |
| Quantity | 5 mg |
| Show More |