Learn More
Alendronic Acid-d6 (Major), TRC
Specifications
Specifications
| Chemical Name or Material | Alendronic Acid-d6 |
| CAS | 1035437-39-8 |
| Molecular Formula | C4 D6 H7 N O7 P2 |
| Quantity | 2.5 mg |
| Synonym | Phosphonic acid, P,P'-(4-amino-1-hydroxybutylidene-2,2,3,3,4,4-d6)bis- (ACI), P,P'-(4-Amino-1-hydroxybutylidene-2,2,3,3,4,4-d6)bis[phosphonic acid] (ACI), 4-Amino-1-hydroxy-1,1-butanediphosphonic-2,2,3,3,4,4-d6 acid, Alendronic Acid-d6, Alendronic Acid-D6 (3-aminopropyl-1,1,2,2,3,3-D6), Alendronic-2,2,3,3,4,4-d6 Acid |
| InChI Formula | InChI=1S/C4H13NO7P2/c5-3-1-2-4(6,13(7,8)9)14(10,11)12/h6H,1-3,5H2,(H2,7,8,9)(H2,10,11,12)/i1D2,2D2,3D2 |
| SMILES | [2H]C([2H])(N)C([2H])([2H])C([2H])([2H])C(O)(P(=O)(O)O)P(=O)(O)O |
| IUPAC Name | (4-amino-2,2,3,3,4,4-hexadeuterio-1-hydroxy-1-phosphonobutyl)phosphonic acid |
| Molecular Weight (g/mol) | 255.133 |
| Formula Weight | 255.054 |