Learn More
ALDH1A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13951-1-AP-150UL
 View more versions of this product
Product Code. 16842394
  • 4836.10 NOK / 150µL
Description
This protein belongs to the aldehyde dehydrogenase family of proteins. The product of this gene is an enzyme that catalyzes the synthesis of retinoic acid (RA) from retinaldehyde. Retinoic acid, the active derivative of vitamin A (retinol), is a hormonal signaling molecule that functions in developing and adult tissues. The studies of a similar mouse gene suggest that this enzyme and the cytochrome CYP26A1, concurrently establish local embryonic retinoic acid levels which facilitate posterior organ development and prevent spina bifida. Three transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
Specifications
ALDH1A2
Polyclonal
Unconjugated
ALDH1A2
ALDH1A2, RALDH 2, RALDH(II), RALDH2, RALDH2 T, Retinal dehydrogenase 2
Rabbit
Antigen Affinity Chromatography
RUO
116676, 19378, 8854
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O94788, Q62148, Q63639
ALDH1A2
ALDH1A2 Fusion Protein Ag5062
150 μL
Primary
Human, Mouse, Rat