Learn More
Aldehyde dehydrogenase, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25U
Unit Size:
25 units
Specifications
Specifications
| Color | Tan to White |
| CAS | 9028-86-8 |
| Quantity | 25U |
| Product Type | Aldehyde dehydrogenase |
| Form | Powder |