Learn More
VEE GEE Alcohol Hydrometer
Product Code. 17896563 Shop All Thermo Scientific CO₂ Incubators Products
Change view
Click to view available options
Alcohol Range:
0% to 20%
105% to 125%
125% to 145%
145% to 165%
165% to 185%
185 to 206%
20 to 40%
40 to 60%
60 to 80%
75% to 95%
80 to 120%
90 to 110%
Increments:
0.2%/±0.2%
0.2%/±0.3%
0.2%/±0.4%
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Alcohol Range Increments unitSize
17896563 75% to 95% 0.2%/±0.3% Each
17866523 165% to 185% 0.2%/±0.2% Each
17896553 60 to 80% 0.2%/±0.3% Each
17826563 0% to 20% 0.2%/±0.3% Each
17896523 80 to 120% 0.2%/±0.3% Each
17836563 40 to 60% 0.2%/±0.4% Each
17806573 125% to 145% 0.2%/±0.3% Each
17886563 90 to 110% 0.2%/±0.3% Each
17886613 105% to 125% 0.2%/±0.3% Each
17886473 185 to 206% 0.2%/±0.2% Each
17876473 145% to 165% 0.2%/±0.3% Each
17836473 20 to 40% 0.2%/±0.4% Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 17896563 Supplier VEE GEE Supplier No. 6613K
Only null left
Add to basket
Add to basket
Verify the alcohol content of your ethyl alcohol-based spirits with our proof scale hydrometer. Tolerances match those laid out by IRS specifications.
  • Alcohol proof range differ depending on model
  • IRS Specification Size differ depending on model
  • li>Calibration for use at 15.5°C/60°F
  • Approx. Length: 305 mm
Specifications
Type Alcohol Hydrometer
Increments 0.2%/±0.3%
Length (Metric) 30.5 cm
Description Alcohol Hydrometer, Proof Scale, IRS Specification, Size K
Material Glass
Temperature of Standardization 15.5°C (60°F)
Alcohol Range 75% to 95%