Learn More
Tork Alcohol Hand Sanitiser Refill for S4 System
Product Code. 30243831 Shop All Tork Products
Change view
Click to view available options
Form:
Gel
Liquid
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Form unitSize
30243831 Liquid Pack of 6
30243863 Gel Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30243831 Supplier Tork Supplier No. 424115
  • ONLINE EXCLUSIVE PRICE
    1335.00 NOK
    1247.57 NOK / Pack of 6
    Save 87.43 NOK
    6% Off
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
Tork Alcohol Hand Sanitiser is an effective hand sanitiser with 80% denatured alcohol.
  • Help to prevent the spread of infections - this hand sanitiser is fully virucidal and conforms with EN1500, EN12791 and EN14476.
  • Our sanitiser is dermatologically tested and leaves no sticky residue.
  • Help to support better hygiene - this sealed refill bottle has a single-use pump to reduce cross-contamination risk.
  • Reduce waste with a refill bottle that collapses in on itself as it empties.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hand Sanitizer
Color Clear
Form Liquid
Active Ingredients Alcohol
Certifications/Compliance EN1500, EN12791, EN14476
Certifications/Compliance (Europe) EN1500, EN12791, EN14476
Disposable Limited-use
Packaging Plastic Bottle
Quantity 6/Pk.
Volume (Metric) 1 L