Learn More
Albuterol Dimer Ether, 75%, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Albuterol Dimer Ether |
| CAS | 147663-30-7 |
| Molecular Formula | C26 H40 N2 O5 |
| Quantity | 5 mg |
| Synonym | 3,3'-[oxybis(methylene)]bis[α-[[(1,1-dimethylethyl)amino]methyl]-4-hydroxy-benzenemethanol, Sabutamol dimer, Salbutamol Imp. F (EP) |
| InChI Formula | InChI=1 S/C26H40N2O5/c1-25(2,3)27-13-23(31)17-7-9-21(29)19(11-17)15-33-16-20-12-18(8-10-22(20)30)24(32)14-28-26(4,5)6/h7-12,23-24,27-32 H,13-16H2,1-6H3 |
| SMILES | CC(C)(C)NCC(O)c1ccc(O)c(COCc2cc(ccc2O)C(O)CNC(C)(C)C)c1 |
| IUPAC Name | 4-[2-(tert-butylamino)-1-hydroxyethyl]-2-[[5-[2-(tert-butylamino)-1-hydroxyethyl]-2-hydroxyphenyl]methoxymethyl]phenol |
| Molecular Weight (g/mol) | 460.61 |
| Formula Weight | 460.2937 g/mol |