Learn More
BINDER™ Alarm Output
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB 65/130/260/470/720/1060
KB PRO Series
KBF PRO Series
KBF Series
Unit Size:
Each
Description
- Alarm output
- Zero-voltage
- Accessible via 3-pin clamping sleeve (max. 24V/2.5A)
- Acoustic signal that can be switched off
Specifications
Specifications
| For Use With (Equipment) | KBF Series |
| Material | Copper |
| Type | Analog Output |