Includes
column, mounts
Requires
column, mounts
- For use with all OHAUS™ Bench Scales and Indicators
- Can be used with manual light tower
Order Info
column, mounts
Certifications
column, mounts
Compliance
column, mounts
Warranty
column, mounts
column, mounts
Specifications
Specifications
| Color | Red, Orange, Green |
| Height (Metric) | 243 mm |
| Languages | Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX) |
| Weight (Metric) | 210 g |
| Type | Column Kit |