Learn More
Alanine-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Alanine-d4 |
| CAS | 53795-92-9 |
| Molecular Formula | C3 D4 H3 N O2 |
| Quantity | 250 mg |
| Synonym | DL-Alanine-2,3,3,3-d4, DL-Alanine-d4, (R,S)-Alanine-d4, (±)-2-Aminopropionic Acid-d4, (±)-Alanine-d4, DL-Ala-d4, DL-α-Alanine-d4, DL-α-Aminopropionic Acid-d4, NSC 7602-d4, dl-2-Aminopropanoic Acid-d4, dl-Alanine-d4 |
| InChI Formula | InChI=1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/i1D3,2D |
| SMILES | [2H]C([2H])([2H])C([2H])(N)C(=O)O |
| IUPAC Name | 2-amino-2,3,3,3-tetradeuteriopropanoic acid |
| Molecular Weight (g/mol) | 93.12 |
| Formula Weight | 93.0728 |