Learn More
Al-23 Tube (one end closed);OD (mm), 4;ID (mm), 2
Product Code. 11302728
Change view
Click to view available options
Quantity:
100 mm
200 mm
300 mm
600 mm
800 mm
1000 mm
Unit Size:
1000mm
100mm
200mm
300mm
600mm
800mm
Product Code. Quantity unitSize
11302728 600 mm 600mm
11372718 100 mm 100mm
11362718 200 mm 200mm
11382718 300 mm 300mm
11392718 800 mm 800mm
11322728 1000 mm 1000mm
6 options
Product Code. 11302728
Brand: Thermo Scientific Alfa Aesar 030062.EH
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Tube
Name Note One end closed
Quantity 600 mm
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.