Learn More
Al-23 Tube, One End Closed, O.D.: 12mm, I.D.: 8mm, Thermo Scientific Chemicals
Product Code. p-4905348
Change view
Click to view available options
Quantity:
100 mm
200 mm
300 mm
400 mm
600 mm
800 mm
1000 mm
Unit Size:
1000mm
100mm
200mm
300mm
400mm
600mm
800mm
Product Code. Quantity unitSize
11342768 100 mm 100mm
11302768 200 mm 200mm
11322768 300 mm 300mm
15451147 400 mm 400mm
11312768 600 mm 600mm
11382758 800 mm 800mm
11332768 1000 mm 1000mm
7 options
Product Code. 11342768
Brand: Thermo Scientific Alfa Aesar 030073.DM
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Tube
Name Note One end closed
Quantity 100 mm